Methyl 3-iodopyrazine-2-carboxylate 97%

Lieferant: Apollo Scientific
Achtung

OR321370-1G
APOSOR321370-1GEA 109 EUR
APOSOR321370-1G
Methyl 3-iodopyrazine-2-carboxylate 97%
Methyl 3-iodopyrazine-2-carboxylate
Formel: C₆H₅IN₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD13195309
CAS-Nummer: 173290-17-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR