4-Azaindole-3-carboxylic acid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1H-Pyrrolo[3,2-b]pyridine-3-carboxylic acid

OR321374-1G
APOSOR321374-1GEA 85.7 EUR
APOSOR321374-1G
4-Azaindole-3-carboxylic acid 97%
4-Azaindole-3-carboxylic acid
Formel: C₈H₆N₂O₂
Molecular Weight: 162,1454 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD11559014
CAS-Nummer: 860496-20-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR