Ethyl-1H-pyrrolo[3,2-c]pyridine-2-carboxylate 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl 1H-pyrrolo[3,2-c]pyridine-2-carboxylate

OR321375-1G
APOSOR321375-1GEA 190 EUR
APOSOR321375-1G
Ethyl-1H-pyrrolo[3,2-c]pyridine-2-carboxylate 97%
Ethyl-1H-pyrrolo[3,2-c]pyridine-2-carboxylate
Formel: C₁₀H₁₀N₂O₂
Molecular Weight: 190,2000 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09878684
CAS-Nummer: 800401-64-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR