Ethyl 3-chloroimidazo[1,2-a]pyrimidine-2-carboxylate, tech

Lieferant: Apollo Scientific

OR32138-1G
APOSOR32138-1GEA 1570 EUR
APOSOR32138-1G
Ethyl 3-chloroimidazo[1,2-a]pyrimidine-2-carboxylate, tech
Ethyl 3-chloroimidazo[1,2-a]pyrimidine-2-carboxylate
Formel: C₉H₈ClN₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD04124241
CAS-Nummer: 860612-52-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR