(R)-2-Benzylpiperidine hydrochloride

Lieferant: Apollo Scientific

OR321380-1G OR321380-250MG
APOSOR321380-1GEA 0 EUR
APOSOR321380-1G APOSOR321380-250MG
(R)-2-Benzylpiperidine hydrochloride
(R)-2-Benzylpiperidine hydrochloride
Formel: C₁₂H₁₇N·ClH
Lagertemperatur: Kühlschrank
MDL: MFCD30728432
CAS-Nummer: 2055172-15-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR