(S)-2-Benzylpiperidine hydrochloride

Lieferant: Apollo Scientific

OR321381-1G OR321381-250MG
APOSOR321381-1GEA 638 EUR
APOSOR321381-1G APOSOR321381-250MG
(S)-2-Benzylpiperidine hydrochloride
(S)-2-Benzylpiperidine hydrochloride
Formel: C₁₂H₁₇N·ClH
Molecular Weight: 211.731 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD30184774
CAS-Nummer: 2055172-15-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR