2-[4-(4-tert-Butylphenyl)-3-methyl-5-oxo-4,5-dihydro-1H-1,2,4-triazol-1-yl]acetonitrile, tech

Lieferant: Apollo Scientific

OR32139-1G
APOSOR32139-1GEA 1570 EUR
APOSOR32139-1G
2-[4-(4-tert-Butylphenyl)-3-methyl-5-oxo-4,5-dihydro-1H-1,2,4-triazol-1-yl]acetonitrile, tech
2-[4-(4-tert-Butylphenyl)-3-methyl-5-oxo-4,5-dihydro-1H-1,2,4-triazol-1-yl]acetonitrile
Formel: C₁₅H₁₈N₄O
Molecular Weight: 270,33 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03848504
CAS-Nummer: 860612-55-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR