N-(3-(Piperidin-4-yl)phenyl)acetamide hydrochloride

Lieferant: Apollo Scientific

OR321390-1G OR321390-250MG
APOSOR321390-1GEA 877 EUR
APOSOR321390-1G APOSOR321390-250MG
N-(3-(Piperidin-4-yl)phenyl)acetamide hydrochloride
N-(3-(Piperidin-4-yl)phenyl)acetamide hydrochloride
Formel: C₁₃H₁₈N₂O·ClH
Lagertemperatur: Kühlschrank
MDL: MFCD18071807
CAS-Nummer: 486430-92-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR