2-Cyan-4,6-dichloranilin 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR3214-1G
APOSOR3214-1GEA
21.9
EUR
APOSOR3214-1G
2-Cyan-4,6-dichloranilin 98%
2-Cyan-4,6-dichloranilin
Formel:
C₇H₄Cl₂N₂ Molecular Weight: 187,03 g/mol Schmelzpunkt: 123…127 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00158947 CAS-Nummer: 36764-94-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...