4-[(6-Chloropyridin-3-yl)methoxy]-3-methoxybenzaldehyde, tech
Lieferant: Apollo Scientific
OR32141-1G
APOSOR32141-1GEA
1570
EUR
APOSOR32141-1G
4-[(6-Chloropyridin-3-yl)methoxy]-3-methoxybenzaldehyde, tech
4-[(6-Chloropyridin-3-yl)methoxy]-3-methoxybenzaldehyde
Formel:
C₁₄H₁₂ClNO₃ Molecular Weight: 277,703 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04124254 CAS-Nummer: 860644-64-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...