N-Methyl-3-(piperidin-4-yl)benzamide hydrochloride

Lieferant: Apollo Scientific

OR321412-1G OR321412-250MG
APOSOR321412-1GEA 0 EUR
APOSOR321412-1G APOSOR321412-250MG
N-Methyl-3-(piperidin-4-yl)benzamide hydrochloride
N-Methyl-3-(piperidin-4-yl)benzamide hydrochloride
Formel: C₁₃H₁₈N₂O·2ClH
Lagertemperatur: Kühlschrank
MDL: MFCD29056884
CAS-Nummer: 1221279-03-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR