4-(Piperidin-4-yl)isoquinoline dihydrochloride

Lieferant: Apollo Scientific

OR321413-1G OR321413-250MG
APOSOR321413-1GEA 647 EUR
APOSOR321413-1G APOSOR321413-250MG
4-(Piperidin-4-yl)isoquinoline dihydrochloride
4-(Piperidin-4-yl)isoquinoline dihydrochloride
Formel: C₁₄H₁₆N₂·2ClH
Molecular Weight: 285.212 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD31629927
CAS-Nummer: 2244087-11-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR