4-(Piperidin-4-yl)isoquinoline dihydrochloride
Lieferant: Apollo Scientific
OR321413-1G
OR321413-250MG
APOSOR321413-1GEA
647
EUR
APOSOR321413-1G
APOSOR321413-250MG
4-(Piperidin-4-yl)isoquinoline dihydrochloride
4-(Piperidin-4-yl)isoquinoline dihydrochloride
Formel:
C₁₄H₁₆N₂·2ClH Molecular Weight: 285.212 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD31629927 CAS-Nummer: 2244087-11-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...