3,5-Dimethyl-1-(4-methylphenyl)-1H-pyrazole-4-carboxylic acid, tech

Lieferant: Apollo Scientific

OR32143-1G
APOSOR32143-1GEA 1570 EUR
APOSOR32143-1G
3,5-Dimethyl-1-(4-methylphenyl)-1H-pyrazole-4-carboxylic acid, tech
3,5-Dimethyl-1-(4-methylphenyl)-1H-pyrazole-4-carboxylic acid
Formel: C₁₃H₁₄N₂O₂
Molecular Weight: 230,262 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05974918
CAS-Nummer: 108444-21-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR