2,2'-((2-((2-Hydroxyethyl)amino)ethyl)azanediyl)bis(ethan-1-ol) 95%

Lieferant: Apollo Scientific

Synonyme: 2,2'-((2-((2-Hydroxyethyl)amino)ethyl)azanediyl)diethanol

OR321430-250MG
APOSOR321430-250MGEA 538 EUR
APOSOR321430-250MG
2,2'-((2-((2-Hydroxyethyl)amino)ethyl)azanediyl)bis(ethan-1-ol) 95%
2,2'-((2-((2-Hydroxyethyl)amino)ethyl)azanediyl)bis(ethan-1-ol)
Formel: C₈H₂₀N₂O₃
Molecular Weight: 192,256 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20667837
CAS-Nummer: 60487-26-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR