2-(1-Methylhydrazin-1-yl)pyrazine 95%

Lieferant: Apollo Scientific

OR32144-1G
APOSOR32144-1GEA 671 EUR
APOSOR32144-1G
2-(1-Methylhydrazin-1-yl)pyrazine 95%
2-(1-Methylhydrazin-1-yl)pyrazine
Formel: C₅H₈N₄
Molecular Weight: 124,144 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05975001
CAS-Nummer: 76319-95-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR