(S)-2-(1-(tert-Butoxycarbonyl)piperidin-3-yl)acetic acid 95%
Lieferant: Apollo Scientific
OR321448-250MG
APOSOR321448-250MGEA
386
EUR
APOSOR321448-250MG
(S)-2-(1-(tert-Butoxycarbonyl)piperidin-3-yl)acetic acid 95%
(S)-2-(1-(tert-Butoxycarbonyl)piperidin-3-yl)acetic acid
Formel:
C₁₂H₂₁NO₄ Molecular Weight: 243,299 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03094722 CAS-Nummer: 941289-27-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...