Methyl 1-(pyrazin-2-yl)piperidine-4-carboxylate, tech

Lieferant: Apollo Scientific

OR32145-1G
APOSOR32145-1GEA 1570 EUR
APOSOR32145-1G
Methyl 1-(pyrazin-2-yl)piperidine-4-carboxylate, tech
Methyl 1-(pyrazin-2-yl)piperidine-4-carboxylate
Formel: C₁₁H₁₅N₃O₂
Molecular Weight: 221,256 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05975002
CAS-Nummer: 860648-97-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR