tert-Butyl-4-((methylamino)methyl)piperidine-1-carboxylate 97%

Lieferant: Apollo Scientific

Synonyme: tert-Butyl 4-((methylamino)methyl)piperidine-1-carboxylate

OR321451-5G
APOSOR321451-5GEA 104 EUR
APOSOR321451-5G
tert-Butyl-4-((methylamino)methyl)piperidine-1-carboxylate 97%
tert-Butyl-4-((methylamino)methyl)piperidine-1-carboxylate
Formel: C₁₂H₂₄N₂O₂
Molecular Weight: 228,3300 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06656629
CAS-Nummer: 138022-02-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR