1-Benzyl-3-bromopyrrolidine
Lieferant: Apollo Scientific
OR321460-1G
OR321460-250MG
OR321460-100MG
APOSOR321460-1GEA
1450
EUR
APOSOR321460-1G
APOSOR321460-250MG
APOSOR321460-100MG
1-Benzyl-3-bromopyrrolidine
1-Benzyl-3-bromopyrrolidine
Formel:
C₁₁H₁₄BrN Molecular Weight: 240.14 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD11226925 CAS-Nummer: 38042-74-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...