3-Acetyl-1-ethyl-4-hydroxy-1,2-dihydroquinolin-2-one 95%
Lieferant: Apollo Scientific
OR32147-1G
APOSOR32147-1GEA
874
EUR
APOSOR32147-1G
3-Acetyl-1-ethyl-4-hydroxy-1,2-dihydroquinolin-2-one 95%
3-Acetyl-1-ethyl-4-hydroxy-1,2-dihydroquinolin-2-one
Formel:
C₁₃H₁₃NO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00226605 CAS-Nummer: 66134-57-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...