2-(Tetrahydrofuran-3-yl)acetic acid 97%

Lieferant: Apollo Scientific

OR321471-1G OR321471-5G
APOSOR321471-1GEA 0 EUR
APOSOR321471-1G APOSOR321471-5G
2-(Tetrahydrofuran-3-yl)acetic acid 97%
2-(Tetrahydrofuran-3-yl)acetic acid
Formel: C₆H₁₀O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD06411265
CAS-Nummer: 138498-97-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR