3-Acetyl-4-hydroxy-1-methyl-1,2-dihydroquinolin-2-one 95%
Lieferant: Apollo Scientific
OR32148-1G
APOSOR32148-1GEA
874
EUR
APOSOR32148-1G
3-Acetyl-4-hydroxy-1-methyl-1,2-dihydroquinolin-2-one 95%
3-Acetyl-4-hydroxy-1-methyl-1,2-dihydroquinolin-2-one
Formel:
C₁₂H₁₁NO₃ Molecular Weight: 217,221 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00220073 CAS-Nummer: 54289-76-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...