7-Chloro-1H-pyrrolo[3,2-b]pyridine 97%

Lieferant: Apollo Scientific

OR321484-5G
APOSOR321484-5GEA 201 EUR
APOSOR321484-5G
7-Chloro-1H-pyrrolo[3,2-b]pyridine 97%
7-Chloro-1H-pyrrolo[3,2-b]pyridine
Formel: C₇H₅ClN₂
Molecular Weight: 152.5800 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08234945
CAS-Nummer: 357263-48-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR