6-Bromo-1H-pyrrolo[2,3-b]pyridine 97%

Lieferant: Apollo Scientific

Synonyme: 6-Bromo-7-azaindole

OR321485-5G
APOSOR321485-5GEA 172 EUR
APOSOR321485-5G
6-Bromo-1H-pyrrolo[2,3-b]pyridine 97%
6-Bromo-1H-pyrrolo[2,3-b]pyridine
Formel: C₇H₅BrN₂
Molecular Weight: 197.04 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06659667
CAS-Nummer: 143468-13-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR