tert-Butyl 4-(4-(aminomethyl)phenyl)piperidine-1-carboxylate
Lieferant: Apollo Scientific
OR321488-250MG
OR321488-500MG
APOSOR321488-250MGEA
171
EUR
APOSOR321488-250MG
APOSOR321488-500MG
tert-Butyl 4-(4-(aminomethyl)phenyl)piperidine-1-carboxylate
tert-Butyl 4-(4-(aminomethyl)phenyl)piperidine-1-carboxylate
Formel:
C₁₇H₂₆N₂O₂ Molecular Weight: 290.401 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD09038246 CAS-Nummer: 1198286-87-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...