3-Acetyl-1-benzyl-4-hydroxy-1,2-dihydroquinolin-2-one, tech

Lieferant: Apollo Scientific

OR32149-1G
APOSOR32149-1GEA 1570 EUR
APOSOR32149-1G
3-Acetyl-1-benzyl-4-hydroxy-1,2-dihydroquinolin-2-one, tech
3-Acetyl-1-benzyl-4-hydroxy-1,2-dihydroquinolin-2-one
Formel: C₁₈H₁₅NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD03230499
CAS-Nummer: 161185-40-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR