4-(3-Ethylphenyl)piperidine hydrochloride

Lieferant: Apollo Scientific

OR321491-1G OR321491-250MG
APOSOR321491-1GEA 493 EUR
APOSOR321491-1G APOSOR321491-250MG
4-(3-Ethylphenyl)piperidine hydrochloride
4-(3-Ethylphenyl)piperidine hydrochloride
Formel: C₁₃H₁₉N·ClH
Lagertemperatur: Kühlschrank
MDL: MFCD30099246
CAS-Nummer: 2256060-53-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR