Ethyl (R)-4-(piperidin-2-yl)benzoate hydrochloride

Lieferant: Apollo Scientific

OR321493-1G OR321493-250MG
APOSOR321493-1GEA 0 EUR
APOSOR321493-1G APOSOR321493-250MG
Ethyl (R)-4-(piperidin-2-yl)benzoate hydrochloride
Ethyl (R)-4-(piperidin-2-yl)benzoate hydrochloride
Formel: C₁₄H₁₉NO₂·ClH
Lagertemperatur: Kühlschrank
MDL: MFCD30738189
CAS-Nummer: 2307737-09-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR