(R)-3-(4-Bromophenyl)piperidine hydrochloride

Lieferant: Apollo Scientific

OR321496-1G OR321496-250MG
APOSOR321496-1GEA 667 EUR
APOSOR321496-1G APOSOR321496-250MG
(R)-3-(4-Bromophenyl)piperidine hydrochloride
(R)-3-(4-Bromophenyl)piperidine hydrochloride
Formel: C₁₁H₁₄BrN·ClH
Lagertemperatur: Kühlschrank
CAS-Nummer: 2409590-07-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR