(1S,3R)-3-(tert-Butoxycarbonylamino)cyclohexanecarboxylic acid

Lieferant: Apollo Scientific

Synonyme: (1S,3R)-3-(tert-Butoxycarbonylamino)cyclohexanecarboxylic acid, (1S,3R)-3-(Boc-amino)cyclohexanecarboxylic Acid, (1S,3R)-3-((tert-butoxycarbonyl)amino)cyclohexanecarboxylic acid

OR321497-250MG
APOSOR321497-250MGEA 30.6 EUR
APOSOR321497-250MG
(1S,3R)-3-(tert-Butoxycarbonylamino)cyclohexanecarboxylic acid
(1S,3R)-3-(tert-Butoxycarbonylamino)cyclohexanecarboxylic acid
Formel: C₁₂H₂₁NO₄
Molecular Weight: 243,30 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD06227743
CAS-Nummer: 222530-34-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR