3-Formyl-2,6-dimethoxypyridine 97%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2,6-Dimethoxynicotinaldehyde, 2,6-Dimethoxypyridine-3-carboxaldehyde

OR3215-1G
APOSOR3215-1GEA 39.3 EUR
APOSOR3215-1G
3-Formyl-2,6-dimethoxypyridine 97%
3-Formyl-2,6-dimethoxypyridine
Formel: C₈H₉NO₃
Molecular Weight: 167,17 g/mol
Schmelzpunkt: 68…72 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08064048
CAS-Nummer: 58819-72-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR