3-(Piperidin-3-yl)benzamide hydrochloride

Lieferant: Apollo Scientific

OR321504-1G OR321504-250MG
APOSOR321504-1GEA 0 EUR
APOSOR321504-1G APOSOR321504-250MG
3-(Piperidin-3-yl)benzamide hydrochloride
3-(Piperidin-3-yl)benzamide hydrochloride
Formel: C₁₂H₁₆N₂O·ClH
Lagertemperatur: Kühlschrank
CAS-Nummer: 2409589-71-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR