3-(Piperidin-3-yl)benzamide hydrochloride
Lieferant: Apollo Scientific
OR321504-1G
OR321504-250MG
APOSOR321504-1GEA
0
EUR
APOSOR321504-1G
APOSOR321504-250MG
3-(Piperidin-3-yl)benzamide hydrochloride
3-(Piperidin-3-yl)benzamide hydrochloride
Formel:
C₁₂H₁₆N₂O·ClH Lagertemperatur: Kühlschrank |
CAS-Nummer:
2409589-71-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...