4-(Piperidin-4-yl)benzonitrile hydrochloride

Lieferant: Apollo Scientific

OR321508-1G OR321508-250MG
APOSOR321508-1GEA 589 EUR
APOSOR321508-1G APOSOR321508-250MG
4-(Piperidin-4-yl)benzonitrile hydrochloride
4-(Piperidin-4-yl)benzonitrile hydrochloride
Formel: C₁₂H₁₄N₂·ClH
Molecular Weight: 222.714 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD08544124
CAS-Nummer: 162997-34-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR