3-Acetyl-4-hydroxy-1-(propan-2-yl)-1,2-dihydroquinolin-2-one, tech

Lieferant: Apollo Scientific

OR32151-1G
APOSOR32151-1GEA 1570 EUR
APOSOR32151-1G
3-Acetyl-4-hydroxy-1-(propan-2-yl)-1,2-dihydroquinolin-2-one, tech
3-Acetyl-4-hydroxy-1-(propan-2-yl)-1,2-dihydroquinolin-2-one
Formel: C₁₄H₁₅NO₃
Molecular Weight: 245,274 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05974941
CAS-Nummer: 860649-18-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR