3-Acetyl-1-butyl-4-hydroxy-1,2-dihydroquinolin-2-one, tech

Lieferant: Apollo Scientific

OR32152-1G
APOSOR32152-1GEA 1570 EUR
APOSOR32152-1G
3-Acetyl-1-butyl-4-hydroxy-1,2-dihydroquinolin-2-one, tech
3-Acetyl-1-butyl-4-hydroxy-1,2-dihydroquinolin-2-one
Formel: C₁₅H₁₇NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00101165
CAS-Nummer: 161185-41-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR