(2R,4S)-N-Boc-4-hydroxypiperidine-2-carboxylic acidmethyl ester

Lieferant: Apollo Scientific

Synonyme: 1-tert-Butyl 2-methyl (2R,4S)-4-hydroxypiperidine-1,2-dicarboxylate

OR321526-250MG
APOSOR321526-250MGEA 114 EUR
APOSOR321526-250MG
(2R,4S)-N-Boc-4-hydroxypiperidine-2-carboxylic acidmethyl ester
(2R,4S)-N-Boc-4-hydroxypiperidine-2-carboxylic acidmethyl ester
Formel: C₁₂H₂₁NO₅
Lagertemperatur: Kühlschrank
MDL: MFCD02259744
CAS-Nummer: 321744-26-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR