3-Acetyl-1-cyclohexyl-4-hydroxy-1,2-dihydroquinolin-2-one, tech

Lieferant: Apollo Scientific

OR32153-1G
APOSOR32153-1GEA 1570 EUR
APOSOR32153-1G
3-Acetyl-1-cyclohexyl-4-hydroxy-1,2-dihydroquinolin-2-one, tech
3-Acetyl-1-cyclohexyl-4-hydroxy-1,2-dihydroquinolin-2-one
Formel: C₁₇H₁₉NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD05974942
CAS-Nummer: 860649-19-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR