Methyl 4-(piperidin-2-yl)benzoate hydrochloride

Lieferant: Apollo Scientific

OR321532-1G OR321532-250MG
APOSOR321532-1GEA 0 EUR
APOSOR321532-1G APOSOR321532-250MG
Methyl 4-(piperidin-2-yl)benzoate hydrochloride
Methyl 4-(piperidin-2-yl)benzoate hydrochloride
Formel: C₁₃H₁₇NO₂·ClH
Lagertemperatur: Kühlschrank
MDL: MFCD02179145
CAS-Nummer: 863769-42-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR