tert-Butyl ((1S,3R)-3-(hydroxymethyl)cyclohexyl)carbamate

Lieferant: Apollo Scientific

OR321534-100MG
APOSOR321534-100MGEA 364 EUR
APOSOR321534-100MG
tert-Butyl ((1S,3R)-3-(hydroxymethyl)cyclohexyl)carbamate
tert-Butyl ((1S,3R)-3-(hydroxymethyl)cyclohexyl)carbamate
Formel: C₁₂H₂₃NO₃
Molecular Weight: 229.316 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD14635493
CAS-Nummer: 1932640-02-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR