tert-Butyl 3-(3-bromophenyl)piperidine-1-carboxylate
Lieferant: Apollo Scientific
OR321535-250MG
APOSOR321535-250MGEA
473
EUR
APOSOR321535-250MG
tert-Butyl 3-(3-bromophenyl)piperidine-1-carboxylate
tert-Butyl 3-(3-bromophenyl)piperidine-1-carboxylate
Formel:
C₁₆H₂₂BrNO₂ Molecular Weight: 340.255 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD11223611 CAS-Nummer: 1203686-41-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...