tert-Butyl 3-(2-bromophenyl)piperidine-1-carboxylate

Lieferant: Apollo Scientific

OR321536-1G OR321536-250MG
APOSOR321536-1GEA 638 EUR
APOSOR321536-1G APOSOR321536-250MG
tert-Butyl 3-(2-bromophenyl)piperidine-1-carboxylate
tert-Butyl 3-(2-bromophenyl)piperidine-1-carboxylate
Formel: C₁₆H₂₂BrNO₂
Molecular Weight: 340.255 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD11223612
CAS-Nummer: 1203683-54-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR