tert-Butyl 3-(2-bromophenyl)piperidine-1-carboxylate
Lieferant: Apollo Scientific
OR321536-1G
OR321536-250MG
APOSOR321536-1GEA
638
EUR
APOSOR321536-1G
APOSOR321536-250MG
tert-Butyl 3-(2-bromophenyl)piperidine-1-carboxylate
tert-Butyl 3-(2-bromophenyl)piperidine-1-carboxylate
Formel:
C₁₆H₂₂BrNO₂ Molecular Weight: 340.255 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD11223612 CAS-Nummer: 1203683-54-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...