Ethyl (S)-2-(piperidin-2-yl)acetate hydrochloride

Lieferant: Apollo Scientific

OR321539-1G OR321539-250MG
APOSOR321539-1GEA 0 EUR
APOSOR321539-1G APOSOR321539-250MG
Ethyl (S)-2-(piperidin-2-yl)acetate hydrochloride
Ethyl (S)-2-(piperidin-2-yl)acetate hydrochloride
Formel: C₉H₁₇NO₂·ClH
Lagertemperatur: Kühlschrank
CAS-Nummer: 2409589-84-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR