Boc-(2S,5R)-5-phenylpyrrolidine-2-carboxylic acid

Lieferant: Apollo Scientific

Synonyme: (2S,5R)-1-(tert-Butoxycarbonyl)-5-phenylpyrrolidine-2-carboxylic acid

OR321545-250MG
APOSOR321545-250MGEA 299 EUR
APOSOR321545-250MG
Boc-(2S,5R)-5-phenylpyrrolidine-2-carboxylic acid
Boc-(2S,5R)-5-phenylpyrrolidine-2-carboxylic acid
Formel: C₁₆H₂₁NO₄
Molecular Weight: 291.35 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD01321008
CAS-Nummer: 221352-49-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR