3-(p-Tolyl)piperidin-3-ol hydrochloride

Lieferant: Apollo Scientific

OR321548-1G OR321548-250MG
APOSOR321548-1GEA 560 EUR
APOSOR321548-1G APOSOR321548-250MG
3-(p-Tolyl)piperidin-3-ol hydrochloride
3-(p-Tolyl)piperidin-3-ol hydrochloride
Formel: C₁₂H₁₇NO·ClH
Lagertemperatur: Kühlschrank
MDL: MFCD28108399
CAS-Nummer: 173447-85-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR