Rel-Methyl (3R,6R)-6-methylpiperidine-3-carboxylate hydrochloride
Lieferant: Apollo Scientific
Synonyme:
trans-Methyl 6-methylpiperidine-3-carboxylate hydrochloride
OR321549-1G
OR321549-250MG
APOSOR321549-1GEA
498
EUR
APOSOR321549-1G
APOSOR321549-250MG
Rel-Methyl (3R,6R)-6-methylpiperidine-3-carboxylate hydrochloride
Rel-Methyl (3R,6R)-6-methylpiperidine-3-carboxylate hydrochloride
Formel:
C₈H₁₅NO₂·ClH Lagertemperatur: Kühlschrank |
MDL:
MFCD30181463 CAS-Nummer: 1009376-86-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...