3-Acetyl-4-hydroxy-1-azatricyclo[7.3.1.05,13]trideca-3,5(13),6,8-tetraen-2-one, tech
Lieferant: Apollo Scientific
OR32155-1G
APOSOR32155-1GEA
1570
EUR
APOSOR32155-1G
3-Acetyl-4-hydroxy-1-azatricyclo[7.3.1.05,13]trideca-3,5(13),6,8-tetraen-2-one, tech
3-Acetyl-4-hydroxy-1-azatricyclo[7.3.1.05,13]trideca-3,5(13),6,8-tetraen-2-one
Formel:
C₁₄H₁₃NO₃ Molecular Weight: 243,258 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02178524 CAS-Nummer: 141945-41-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...