3-(2H-Pyrazol-3-yl)piperidine dihydrochloride

Lieferant: Apollo Scientific

OR321551-250MG
APOSOR321551-250MGEA 134 EUR
APOSOR321551-250MG
3-(2H-Pyrazol-3-yl)piperidine dihydrochloride
3-(2H-Pyrazol-3-yl)piperidine dihydrochloride
Formel: C₈H₁₅Cl₂N₃
Molecular Weight: 224,13 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD11839730
CAS-Nummer: 51747-03-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR